Learn More
Medchemexpress LLC 5-Aminosalicylic acid | 89-57-6 | C7H7NO3 | 50 G
SDP
Catalog No. 5000374000
Encompass_Preferred
5-Aminosalicylic acid (Mesalamine) is a compound that acts as a specific PPARγ agonist and inhibits p21-activated kinase 1 (PAK1) and NF-κB. It also shows an ability to inhibit the activity of osteopontin (OPN). This solid appears brown to gray and is stable under recommended storage conditions, making it suitable for various laboratory applications.
  • Acts as a specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1) and NF-κB
  • Inhibits the activity of osteopontin (OPN)
  • Stable under recommended storage conditions
  • Suitable for laboratory chemicals and manufacturing substances
Catalog No. 50-003-74000 Supplier Medchemexpress LLC Supplier No. HY1502750G
Add to Cart
Add to Cart
5-Aminosalicylic acid (Mesalamine) is a compound that acts as a specific PPARγ agonist and inhibits p21-activated kinase 1 (PAK1) and NF-κB. It also shows an ability to inhibit the activity of osteopontin (OPN). This solid appears brown to gray and is stable under recommended storage conditions, making it suitable for various laboratory applications.
  • Acts as a specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1) and NF-κB
  • Inhibits the activity of osteopontin (OPN)
  • Stable under recommended storage conditions
  • Suitable for laboratory chemicals and manufacturing substances