Learn More
Cayman Chemical 5-AmInosalIcylIc AcId-d7 1mg
SDP
Catalog No. 502919444
Encompass_Preferred
An internal standard for the quantification of 5-aminosalicylic acid by GC- or LC-MS
Catalog No. 50-291-9444 Supplier Cayman Chemical Supplier No. 371971 MG
Add to Cart
Add to Cart
An internal standard for the quantification of 5-aminosalicylic acid by GC- or LC-MS