Learn More
Medchemexpress LLC 5-Aminosalicylic Acid | MFCD00007877 | 25 MG
SDP
Catalog No. 5000374499
Encompass_Preferred
5-Aminosalicylic acid (Mesalamine) acts as a specific PPARγ agonist and also inhibits p21-activated kinase 1 (PAK1) and NF-κB. It can also inhibit the activity of osteopontin (OPN).
  • Specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1)
  • Inhibits NF-κB
  • Inhibits osteopontin (OPN) activity
  • Targets PPAR, PAK, NF-κB, endogenous metabolite
  • Research areas: inflammation/immunology and cancer
Catalog No. 50-003-74499 Supplier Medchemexpress LLC Supplier No. 25MGHY15027R
Add to Cart
Add to Cart
5-Aminosalicylic acid (Mesalamine) acts as a specific PPARγ agonist and also inhibits p21-activated kinase 1 (PAK1) and NF-κB. It can also inhibit the activity of osteopontin (OPN).
  • Specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1)
  • Inhibits NF-κB
  • Inhibits osteopontin (OPN) activity
  • Targets PPAR, PAK, NF-κB, endogenous metabolite
  • Research areas: inflammation/immunology and cancer