Learn More
Medchemexpress LLC 5-Aminosalicylic Acid (Standard) | 89-57-6 | C7H7NO3 | 250 MG
SDP
Catalog No. 5000374486
Encompass_Preferred
5-Aminosalicylic Acid (Standard) is an analytical standard used in research and analytical applications. Also known as Mesalamine, 5-ASA, or Mesalazine, it acts as a specific PPARγ agonist and inhibits p21-activated kinase 1 (PAK1) and NF-κB. This compound can also inhibit the activity of osteopontin (OPN) and is commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Acts as a specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1) and NF-κB
  • Inhibits the activity of osteopontin (OPN)
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-003-74486 Supplier Medchemexpress LLC Supplier No. 250MGHY15027R
Add to Cart
Add to Cart
5-Aminosalicylic Acid (Standard) is an analytical standard used in research and analytical applications. Also known as Mesalamine, 5-ASA, or Mesalazine, it acts as a specific PPARγ agonist and inhibits p21-activated kinase 1 (PAK1) and NF-κB. This compound can also inhibit the activity of osteopontin (OPN) and is commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Acts as a specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1) and NF-κB
  • Inhibits the activity of osteopontin (OPN)
  • Used in HPLC, GC, and MS research experiments