Learn More
Medchemexpress LLC 5-Aminothalidomide | 191732-76-0 | MFCD30188077 | 99.7% | 273.24 g/mol | C13H11N3O4 | 250 MG
SDP
Catalog No. 5000303650
Encompass_Preferred
5-Aminothalidomide is a thalidomide-derived small molecule that serves as an E3 ligase ligand for PROTAC research. It is used to recruit cereblon in the synthesis of PROTAC degraders and related chemical biology applications, and is supplied with analytical documentation confirming high purity.
  • High purity suitable for synthetic chemistry and biochemical assays.
  • Functionalized for conjugation in PROTAC linker chemistry.
  • Documented with certificate of analysis (COA) and safety data sheet (SDS) for quality and safety verification.
  • Available in multiple laboratory-scale sizes to support research workflows.
  • Compatible with common analytical methods such as HPLC for batch verification.
Catalog No. 50-003-03650 Supplier Medchemexpress LLC Supplier No. 250MGHYW023573
5-Aminothalidomide is a thalidomide-derived small molecule that serves as an E3 ligase ligand for PROTAC research. It is used to recruit cereblon in the synthesis of PROTAC degraders and related chemical biology applications, and is supplied with analytical documentation confirming high purity.
  • High purity suitable for synthetic chemistry and biochemical assays.
  • Functionalized for conjugation in PROTAC linker chemistry.
  • Documented with certificate of analysis (COA) and safety data sheet (SDS) for quality and safety verification.
  • Available in multiple laboratory-scale sizes to support research workflows.
  • Compatible with common analytical methods such as HPLC for batch verification.