Learn More
Cayman Chemical 5-Aminothalidomide, Purity: ≥95%, CAS: 191732-76-0, 500 mg
SDP
Catalog No. 502918938
Encompass_Preferred
A derivative of (±)-thalidomide; inhibits the proliferation of EA.hy926 endothelial cells (IC50 = 200 µM); an intermediate in the synthesis of a PROTAC c-MET degrader with antiproliferative activity in c-MET-sensitive cancer cells; an intermediate in the synthesis of a thalidomide derivative that reduces clinical scores in a mouse model of EAE
Catalog No. 50-291-8938 Supplier Cayman Chemical Supplier No. 39514500 MG
A derivative of (±)-thalidomide; inhibits the proliferation of EA.hy926 endothelial cells (IC50 = 200 µM); an intermediate in the synthesis of a PROTAC c-MET degrader with antiproliferative activity in c-MET-sensitive cancer cells; an intermediate in the synthesis of a thalidomide derivative that reduces clinical scores in a mouse model of EAE