Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-AMP-activated protein kinase (AMPK) α2/β1/γ1 heterotrimer protein, human | >90.0% | 50 UG
SDP

Catalog No. 5000294068
Encompass_Preferred

Recombinant human AMPK α2/β1/γ1 heterotrimer protein expressed in Sf9 insect cells and supplied with N-terminal His and GST tags. This purified research reagent is suitable for biochemical characterization and cellular studies and is quality-controlled by reducing SDS-PAGE. Available in microgram pack sizes for assay development.

  • Recombinant human heterotrimer (α2/β1/γ1) for functional assays.
  • Expressed in Sf9 insect cells to support eukaryotic folding.
  • Tagged with N-terminal His and GST for purification and detection.
  • Purity greater than 90.0% as determined by reducing SDS-PAGE.
  • Available in microgram pack sizes suitable for assay development.

Catalog No. 50-002-94068 Supplier Medchemexpress LLC Supplier No. HYP7614250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human AMPK α2/β1/γ1 heterotrimer protein expressed in Sf9 insect cells and supplied with N-terminal His and GST tags. This purified research reagent is suitable for biochemical characterization and cellular studies and is quality-controlled by reducing SDS-PAGE. Available in microgram pack sizes for assay development.

  • Recombinant human heterotrimer (α2/β1/γ1) for functional assays.
  • Expressed in Sf9 insect cells to support eukaryotic folding.
  • Tagged with N-terminal His and GST for purification and detection.
  • Purity greater than 90.0% as determined by reducing SDS-PAGE.
  • Available in microgram pack sizes suitable for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.