Learn More
Biotium 5-(AND-6)-CARBOXY-X-Rhodamine, SUCCINIMI
Catalog No. 501964799
Encompass_Preferred
5(6)-ROX, SE (full name: 5-(and-6)-Carboxy-X-rhodamine, succinimidyl ester) is an amine-reactive long wavelength rhodamine dye. Properties: Ex/Em (MeOH) = 568/595 nm; Dark red solid soluble in DMSO or DMF; Store at -20°C and protect from light; C 37 H 33 N 3 O 7; MW: 631.7; [114616-32-9].
Catalog No. 50-196-4799 Supplier Biotium Supplier No. 90005
Add to Cart
Edge
Add to Cart
5(6)-ROX, SE (full name: 5-(and-6)-Carboxy-X-rhodamine, succinimidyl ester) is an amine-reactive long wavelength rhodamine dye. Properties: Ex/Em (MeOH) = 568/595 nm; Dark red solid soluble in DMSO or DMF; Store at -20°C and protect from light; C 37 H 33 N 3 O 7; MW: 631.7; [114616-32-9].