Learn More
Biotium 5-(AND-6)-CARBOXYTetramethylrhodamine SU
Catalog No. 501964804
Encompass_Preferred
5(6)-TAMRA SE (full name: 5-(and-6)-Carboxytetramethylrhodamine succinimidyl ester (mixed isomers)) is the amine reactive form of TAMRA, one of the most commonly used red fluorescent dyes. The succinimidyl ester reacts readily with primary or secondary amines under mild conditions. Properties: Ex/Em (MeOH) = 540/565 nm; e = 95,000; Red solid soluble in DMF or DMSO; Store at -20°C and protect from light; C 29 H 25 N 3 O 7; MW: 527.5; [150408-83-6]
Catalog No. 50-196-4804 Supplier Biotium Supplier No. 90022
5(6)-TAMRA SE (full name: 5-(and-6)-Carboxytetramethylrhodamine succinimidyl ester (mixed isomers)) is the amine reactive form of TAMRA, one of the most commonly used red fluorescent dyes. The succinimidyl ester reacts readily with primary or secondary amines under mild conditions. Properties: Ex/Em (MeOH) = 540/565 nm; e = 95,000; Red solid soluble in DMF or DMSO; Store at -20°C and protect from light; C 29 H 25 N 3 O 7; MW: 527.5; [150408-83-6]