Learn More
Medchemexpress LLC 5α-Androstenone-d4 | 18339-16-7 | 99.98% | 276.45 | 2.5 MG
SDP
Catalog No. 5000436198
Encompass_Preferred
5α-Androstenone-d4 is a deuterium-labeled form of 5α-Androstenone, used primarily in laboratory research. It serves as a tracer or internal standard for quantitative analysis methods like NMR, GC-MS, and LC-MS. Deuteration has gained attention for its potential to modify the pharmacokinetic and metabolic profiles of drug molecules.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Intended for research use only
  • Not for sale to patients
  • Potential to affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-36198 Supplier Medchemexpress LLC Supplier No. HY118566S2.5MG
5α-Androstenone-d4 is a deuterium-labeled form of 5α-Androstenone, used primarily in laboratory research. It serves as a tracer or internal standard for quantitative analysis methods like NMR, GC-MS, and LC-MS. Deuteration has gained attention for its potential to modify the pharmacokinetic and metabolic profiles of drug molecules.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Intended for research use only
  • Not for sale to patients
  • Potential to affect pharmacokinetic and metabolic profiles