Learn More
Cayman Chemical 5-APBhydrochlorIde 10mg
SDP
Catalog No. 502951555
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; a metabolite of 5-MAPB; a potential metabolite of 5-EAPB; intended for research and forensic applications
Catalog No. 50-295-1555 Supplier Cayman Chemical Supplier No. 1113410 MG
An analytical reference standard categorized as a benzofuran; a metabolite of 5-MAPB; a potential metabolite of 5-EAPB; intended for research and forensic applications