Learn More
Cayman Chemical 5-APDB (hydrochloride), Purity: ≥98%, CAS: 152623-94-4, 10 mg
SDP
Catalog No. 502951144
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; substitutes for (S)-(+)-MBDB and MMAI in two-lever drug discrimination tests in rats; intended for research and forensic applications.
Catalog No. 50-295-1144 Supplier Cayman Chemical Supplier No. 1145610 MG
An analytical reference standard categorized as a benzofuran; substitutes for (S)-(+)-MBDB and MMAI in two-lever drug discrimination tests in rats; intended for research and forensic applications.