Learn More
Cayman Chemical 5-APDIhydrochlorIde 10mg
SDP
Catalog No. 502955292
Encompass_Preferred
An indane with an amphetamine-like aminopropane group; inhibits the uptake of serotonin, dopamine, and norepinephrine by crude synaptosomes (IC50 = 82, 1,847, and 849 nM, respectively)
Catalog No. 50-295-5292 Supplier Cayman Chemical Supplier No. 1389910 MG
An indane with an amphetamine-like aminopropane group; inhibits the uptake of serotonin, dopamine, and norepinephrine by crude synaptosomes (IC50 = 82, 1,847, and 849 nM, respectively)