Learn More
Abcam 5-Aza-2'-deo x ycytidine (Decitabine), DNMT1 inhibitor, 25MG
Catalog No. 502584563
Encompass_Preferred
MW 228.21 Da, Purity >99%. Potent DNA methyltransferase 1 (DNMT1) inhibitor (IC₅₀ = 0.2 μM). Incorporated into DNA, inhibits DNMT1 activity to induce DNA hypomethylation. Interferes with protein synthesis. Shows antimetabolic and anticancer activities in vitro. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4563 Supplier Abcam Supplier No. AB12084225MG
MW 228.21 Da, Purity >99%. Potent DNA methyltransferase 1 (DNMT1) inhibitor (IC₅₀ = 0.2 μM). Incorporated into DNA, inhibits DNMT1 activity to induce DNA hypomethylation. Interferes with protein synthesis. Shows antimetabolic and anticancer activities in vitro. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement