Learn More
Abcam 5-Aza-2'-deo x ycytidine (Decitabine), DNMT1 inhibitor, 10MG
Catalog No. 502584564
Encompass_Preferred
MW 228.21 Da, Purity >99%. Potent DNA methyltransferase 1 (DNMT1) inhibitor (IC₅₀ = 0.2 μM). Incorporated into DNA, inhibits DNMT1 activity to induce DNA hypomethylation. Interferes with protein synthesis. Shows antimetabolic and anticancer activities in vitro. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4564 Supplier Abcam Supplier No. AB12084210MG
MW 228.21 Da, Purity >99%. Potent DNA methyltransferase 1 (DNMT1) inhibitor (IC₅₀ = 0.2 μM). Incorporated into DNA, inhibits DNMT1 activity to induce DNA hypomethylation. Interferes with protein synthesis. Shows antimetabolic and anticancer activities in vitro. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement