Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aza-2'-deoxycytidine, MP Biomedicals™

Catalog No. ICN15480310 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15480310 10 mg
ICN15480305 5 mg
ICN15480325 25 mg
ICN15480350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15480310 Supplier MP Biomedicals, Inc Supplier No. 0215480310
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2353-33-5
Molecular Formula C9H13N3O4
Molecular Weight (g/mol) 227.22
MDL Number MFCD00006547
InChI Key CKTSBUTUHBMZGZ-SHYZEUOFSA-N
Synonym decitabine, 5-aza-2'-deoxycytidine, dacogen, 5-azadeoxycytidine, 2'-deoxy-5-azacytidine, azadc, dezocitidine, 5-aza-cdr, 5-aza-dc, dacogen tn
PubChem CID 451668
ChEBI CHEBI:50131
SMILES NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Color White
Assay Percent Range 99.1%
Quantity 10 mg
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Formula Weight 228.2
Physical Form Crystalline Powder
Chemical Name or Material 5-Aza-2'-deoxycytidine, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.