Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aza-2'-deoxycytidine, TCI America™
SDP

Catalog No. A223220MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A223220MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A223220MG Supplier TCI America Supplier No. A223220MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2353-33-5
Molecular Formula C9H13N3O4
Molecular Weight (g/mol) 227.22
MDL Number MFCD00006547
InChI Key CKTSBUTUHBMZGZ-SHYZEUOFSA-N
Synonym decitabine, 5-aza-2'-deoxycytidine, dacogen, 5-azadeoxycytidine, 2'-deoxy-5-azacytidine, azadc, dezocitidine, 5-aza-cdr, 5-aza-dc, dacogen tn
PubChem CID 451668
ChEBI CHEBI:50131
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Melting Point 191°C
Color White
Quantity 20 mg
Formula Weight 228.21
Physical Form Crystalline Powder
Chemical Name or Material 5-Aza-2′-deoxycytidine
RTECSNumber XZ3012000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.