Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-aza-4'-thio-2'-deoxycytidine | 169514-76-5 | 99.1% | C8H12N4O3S | 10MG
SDP

Catalog No. 5000260644
Encompass_Preferred

5-Aza-4'-thio-2'-deoxycytidine is a sulfur-containing deoxycytidine analog and DNA methyltransferase inhibitor used in preclinical research to induce DNA hypomethylation and evaluate antitumor activity. It is supplied as a high-purity solid suitable for laboratory studies and requires low-temperature storage for stability.

  • High purity (99.09%) for research applications.
  • White to off-white solid appearance.
  • Molecular formula C8H12N4O3S; molecular weight 244.27 g/mol.
  • Powder stable at -20°C for long-term storage.
  • In-solution storage: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-60644 Supplier Medchemexpress LLC Supplier No. HY13901510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Aza-4'-thio-2'-deoxycytidine is a sulfur-containing deoxycytidine analog and DNA methyltransferase inhibitor used in preclinical research to induce DNA hypomethylation and evaluate antitumor activity. It is supplied as a high-purity solid suitable for laboratory studies and requires low-temperature storage for stability.

  • High purity (99.09%) for research applications.
  • White to off-white solid appearance.
  • Molecular formula C8H12N4O3S; molecular weight 244.27 g/mol.
  • Powder stable at -20°C for long-term storage.
  • In-solution storage: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.