Learn More
Cayman Chemical 5-Azacytidine, Purity: ≥95%, CAS: 320-67-2, 50 mg
SDP
Catalog No. 502954978
Encompass_Preferred
An inhibitor of DNA methyltransferases that reduces hypermethylation associated with certain diseases, including myelodysplastic syndromes (IC50s = 2.4 and 2.6 μM for in vitro anti-myeloma activity) and cancer (IC50s ~ 0.4 μM for inhibiting proliferation of various cancer cell lines)
Catalog No. 50-295-4978 Supplier Cayman Chemical Supplier No. 1116450 MG
An inhibitor of DNA methyltransferases that reduces hypermethylation associated with certain diseases, including myelodysplastic syndromes (IC50s = 2.4 and 2.6 μM for in vitro anti-myeloma activity) and cancer (IC50s ~ 0.4 μM for inhibiting proliferation of various cancer cell lines)