Learn More
AdipoGen 5-Azacytidine
SDP
Supplier:  AdipoGen CDXA0271M250
Encompass_Preferred
Chemical. CAS 320-67-2. Formula C8H12N4O5. MW 244.2. Synthetic. 5-Azacytidine, a chemical analogue of the DNA and RNA nucleoside cytidine, is an cell permeable inhibitor of DNA methyltransferases, potentially serving to reverse epigenetic changes. It reduces hypermethylation associated with certain diseases, including myelodysplastic syndromes IC50s = 2.4 and 2.6µM for in vitro anti-myeloma activity and cancer IC50s ~ 0.4µM for inhibiting proliferation of various cancer cell lines. It has a reported half-life of 17 hours and is considerably cytotoxic it must be incorporated into DNA to covalently trap DNA methyltransferases. Induces demethylation and reactivation of silenced genes. Improves the efficiency of reprogramming of stem cells induces differentiation of mesenchymal stem cells into cardiomyocytes.
Catalog No. 50-256-1771