Learn More
Medchemexpress LLC HY-10586 500mg Medchemexpress, 5-Azacytidine Ladakamycin;5-AzaC;Azacitidine CAS:320-67-2 Purity:98%
SDP
Catalog No. 501637297
Encompass_Preferred
Medchemexpress,HY-10586 500mg 5-Azacytidine Ladakamycin;5-AzaC;Azacitidine CAS:320-67-2 Formula:C8H12N4O5 5-Azacytidine is a nucleoside analogue of cytidine that specifically inhibits DNA methylation by trapping DNA methyltransferases. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7297 Supplier Medchemexpress LLC Supplier No. HY10586500MG
Medchemexpress,HY-10586 500mg 5-Azacytidine Ladakamycin;5-AzaC;Azacitidine CAS:320-67-2 Formula:C8H12N4O5 5-Azacytidine is a nucleoside analogue of cytidine that specifically inhibits DNA methylation by trapping DNA methyltransferases. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.