Learn More
TARGETMOL CHEMICALS INC 5-Azacytidine 500MG
SDP
Catalog No. 502875983
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 5-Azacytidine (Ladakamycin) is a cytidine nucleoside analog, a DNA methylation inhibitor with specificity. 5-Azacytidine regulates gene expression by decreasing the level of DNA methylation. 5-Azacytidine induces autophagy and has antitumor activity. Purity 99.62%
Catalog No. 50-287-5983 Supplier TARGETMOL CHEMICALS INC Supplier No. T1339500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 5-Azacytidine (Ladakamycin) is a cytidine nucleoside analog, a DNA methylation inhibitor with specificity. 5-Azacytidine regulates gene expression by decreasing the level of DNA methylation. 5-Azacytidine induces autophagy and has antitumor activity. Purity 99.62%