Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Azacytidine, MP Biomedicals™

Catalog No. ICN10082180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10082180 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10082180 Supplier MP Biomedicals, Inc Supplier No. 0210082180

Chemical Identifiers

CAS 320-67-2
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.21
InChI Key NMUSYJAQQFHJEW-KVTDHHQDSA-N
Synonym 5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin
PubChem CID 9444
ChEBI CHEBI:2038
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydro-1,3,5-triazin-2-one
SMILES NC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 0.1 g
Formula Weight 244.2
Physical Form Powder
Chemical Name or Material 5-Azacytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.