Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Azacytosine

Catalog No. 8550495G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-504-95G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 85-504-95G Supplier Sigma Aldrich Supplier No. 8550495G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5-Azacytosine

Specifications

CAS 931-86-2
Melting Point >300°C (lit.)
Molecular Formula C3H4N4O
Linear Formula C3H4N4O
MDL Number MFCD00006033
Quantity 1 g
Synonym 2-Hydroxy-4-aminotriazine; 4-Amino-1,3,5-triazin-2-ol; 4-Amino-1,3,5-triazin-2-one; 4-Amino-2-hydroxy-1,3,5-triazine; NSC 51100; NSC 54006
Molecular Weight (g/mol) 112.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.