Learn More
Medchemexpress LLC 5-Azaindole | 271-34-1 | 118.14 | 10 G
SDP
Catalog No. 5000449034
Encompass_Preferred
5-Azaindole (1H-Pyrrolo[3,2-c]pyridine) is a biochemical reagent used in life science research. It functions as a factor VIIa inhibitor and is also a valuable intermediate for various pharmaceutical applications. It appears as a solid, ranging from white to yellow.
  • Biochemical reagent for life science research
  • Functions as a factor VIIa inhibitor
  • Useful intermediate for pharmaceutical applications
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-49034 Supplier Medchemexpress LLC Supplier No. HYW00271110G
5-Azaindole (1H-Pyrrolo[3,2-c]pyridine) is a biochemical reagent used in life science research. It functions as a factor VIIa inhibitor and is also a valuable intermediate for various pharmaceutical applications. It appears as a solid, ranging from white to yellow.
  • Biochemical reagent for life science research
  • Functions as a factor VIIa inhibitor
  • Useful intermediate for pharmaceutical applications
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year