Learn More
Medchemexpress LLC 5-azido-PEG4-SS-UTP tristriethylamine | 99.1% | 1279.34 | C46H93N10O21P3S2 | 10 MG
SDP
Catalog No. 5000288652
Encompass_Preferred
5-Azido-PEG4-SS-UTP (tristriethylamine) is an azide-functionalized uridine triphosphate modified with a PEG4 spacer and a cleavable disulfide linker, supplied as the tristriethylamine salt for biochemical labeling applications. It is intended for enzymatic incorporation into RNA and for bioorthogonal click chemistry to attach probes or affinity tags.
  • Azide-functionalized UTP with PEG4 spacer and disulfide linker.
  • Enzymatic incorporation into RNA for site-specific labeling.
  • Provides an azide handle compatible with click chemistry.
  • Supplied as a tristriethylamine salt to aid handling and solubility.
  • Recommended storage: 4°C sealed; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small laboratory quantities (1 mg-25 mg) for research use.
Catalog No. 50-002-88652 Supplier Medchemexpress LLC Supplier No. HY15838510MG
5-Azido-PEG4-SS-UTP (tristriethylamine) is an azide-functionalized uridine triphosphate modified with a PEG4 spacer and a cleavable disulfide linker, supplied as the tristriethylamine salt for biochemical labeling applications. It is intended for enzymatic incorporation into RNA and for bioorthogonal click chemistry to attach probes or affinity tags.
  • Azide-functionalized UTP with PEG4 spacer and disulfide linker.
  • Enzymatic incorporation into RNA for site-specific labeling.
  • Provides an azide handle compatible with click chemistry.
  • Supplied as a tristriethylamine salt to aid handling and solubility.
  • Recommended storage: 4°C sealed; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small laboratory quantities (1 mg-25 mg) for research use.