Learn More
Medchemexpress LLC 5-Azidomethyl-2'-deoxyuridine | 59090-48-1 | C10H13N5O5 | 25 MG
SDP
Catalog No. 5000375219
Encompass_Preferred
5-Azidomethyl-2'-deoxyuridine (5-AmdU) is a purine nucleoside analog and click chemistry reagent, containing an azide group. It demonstrates effective radiosensitization in EMT6 tumor cells and is utilized in various chemical reactions.
  • Functions as a purine nucleoside analog.
  • Demonstrates effective radiosensitization in EMT6 tumor cells.
  • Contains an azide group for click chemistry applications.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne-containing molecules.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
Catalog No. 50-003-75219 Supplier Medchemexpress LLC Supplier No. 25MGHY154253
Add to Cart
Add to Cart
5-Azidomethyl-2'-deoxyuridine (5-AmdU) is a purine nucleoside analog and click chemistry reagent, containing an azide group. It demonstrates effective radiosensitization in EMT6 tumor cells and is utilized in various chemical reactions.
  • Functions as a purine nucleoside analog.
  • Demonstrates effective radiosensitization in EMT6 tumor cells.
  • Contains an azide group for click chemistry applications.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne-containing molecules.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.