Learn More
Medchemexpress LLC 5-Azidopentanoic acid | 79583-98-5 | 99.77% | 25 MG
SDP
Catalog No. 5000341746
Encompass_Preferred
5-Azidopentanoic acid (also known as 5-Azidovaleric acid) is an organic acid primarily utilized in click chemistry reactions. It is intended for research use only and is not sold for patient administration.
  • Organic acid
  • Suitable for click chemistry reactions
  • For research use only
  • Categorized as a biochemical assay reagent
  • Purity of 99.77%
  • Light yellow to brown liquid appearance
Catalog No. 50-003-41746 Supplier Medchemexpress LLC Supplier No. 25MGHY130544
5-Azidopentanoic acid (also known as 5-Azidovaleric acid) is an organic acid primarily utilized in click chemistry reactions. It is intended for research use only and is not sold for patient administration.
  • Organic acid
  • Suitable for click chemistry reactions
  • For research use only
  • Categorized as a biochemical assay reagent
  • Purity of 99.77%
  • Light yellow to brown liquid appearance