Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Azidopentanoic acid | 79583-98-5 | 99.8% | 250 MG
SDP

Catalog No. 5000341456
Encompass_Preferred

5-Azidopentanoic acid (5-Azidovaleric acid) is an organic acid that can be used in click chemistry reactions. It is for research use only and not sold to patients.

  • Molecular weight: 143.14
  • Formula: C₅H₉N₃O₂
  • Appearance: Liquid (density: 1.142 g/cm³)
  • Color: Light yellow to brown
  • Solubility in DMSO: 100 mg/mL (698.62 mM)

Catalog No. 50-003-41456 Supplier Medchemexpress LLC Supplier No. 250MGHY130544
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Azidopentanoic acid (5-Azidovaleric acid) is an organic acid that can be used in click chemistry reactions. It is for research use only and not sold to patients.

  • Molecular weight: 143.14
  • Formula: C₅H₉N₃O₂
  • Appearance: Liquid (density: 1.142 g/cm³)
  • Color: Light yellow to brown
  • Solubility in DMSO: 100 mg/mL (698.62 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.