Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Azidopentanoic acid | 79583-98-5 | 99.8% | 50 MG
SDP

Catalog No. 5000340241
Encompass_Preferred

5-Azidopentanoic acid, also known as 5-Azidovaleric acid, is an organic acid utilized in click chemistry reactions.

  • Also known as 5-azidovaleric acid
  • Organic acid
  • Used in click chemistry reactions
  • Molecular weight: 143.14
  • Chemical formula: C5H9N3O2
  • Appearance: Liquid, light yellow to brown, with a density of 1.142 g/cm3
  • Soluble in DMSO at 100 mg/mL (698.62 mM), requiring sonication
  • Classified under click chemistry, azide, and biochemical assay reagents

Catalog No. 50-003-40241 Supplier Medchemexpress LLC Supplier No. HY13054450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Azidopentanoic acid, also known as 5-Azidovaleric acid, is an organic acid utilized in click chemistry reactions.

  • Also known as 5-azidovaleric acid
  • Organic acid
  • Used in click chemistry reactions
  • Molecular weight: 143.14
  • Chemical formula: C5H9N3O2
  • Appearance: Liquid, light yellow to brown, with a density of 1.142 g/cm3
  • Soluble in DMSO at 100 mg/mL (698.62 mM), requiring sonication
  • Classified under click chemistry, azide, and biochemical assay reagents

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.