Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Azoniaspiro[4.4]nonane Iodide 98.0+%, TCI America™
SDP

Catalog No. A30935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30935G 5 g
A30931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30935G Supplier TCI America Supplier No. A30935G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 45650-35-9
Molecular Formula C8H16IN
Molecular Weight (g/mol) 253.127
InChI Key DWOWCUCDJIERQX-UHFFFAOYSA-M
PubChem CID 86209376
IUPAC Name 5-azoniaspiro[4.4]nonane;iodide
SMILES C1CC[N+]2(C1)CCCC2.[I-]

Specifications

Color White
Quantity 5 g
Formula Weight 253.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Azoniaspiro[4.4]nonane Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.