Learn More
Medchemexpress LLC 5-BDBD | 768404-03-1 | MFCD18086878 | 99.9% | 355.19 | C17H11BrN2O2 | 50 MG
SDP
Catalog No. 5000248319
Encompass_Preferred
5-BDBD is a selective small-molecule antagonist of the P2X4 receptor used as a research tool in pharmacology and neuroscience. It inhibits P2X4-mediated currents (reported IC50 ≈0.75 μM) and has demonstrated efficacy in electrophysiology and animal pain models when formulated appropriately.
  • Selective P2X4 receptor antagonist for receptor pharmacology studies.
  • Demonstrated activity in electrophysiology and in vivo pain models.
  • High chemical purity (≈99.9%).
  • Available as solid and as 10 mM solution in DMSO for convenience.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Suitable for in vitro assays and in vivo studies with proper formulation.
Catalog No. 50-002-48319 Supplier Medchemexpress LLC Supplier No. HY10191150MG
5-BDBD is a selective small-molecule antagonist of the P2X4 receptor used as a research tool in pharmacology and neuroscience. It inhibits P2X4-mediated currents (reported IC50 ≈0.75 μM) and has demonstrated efficacy in electrophysiology and animal pain models when formulated appropriately.
  • Selective P2X4 receptor antagonist for receptor pharmacology studies.
  • Demonstrated activity in electrophysiology and in vivo pain models.
  • High chemical purity (≈99.9%).
  • Available as solid and as 10 mM solution in DMSO for convenience.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Suitable for in vitro assays and in vivo studies with proper formulation.