Learn More
Chemgenes Corporation 5 Bead Linker
SDP
Catalog No. NC1513163
Encompass
Beads are either available pre-filtered with a size range of 20-40 um, or unfiltered with a larger size range. The beads will arrive as dry resin.
Catalog No. NC1513163 Supplier Chemgenes Corporation Supplier No. MACOSKO201110(V+)
Beads are either available pre-filtered with a size range of 20-40 um, or unfiltered with a larger size range. The beads will arrive as dry resin.