Learn More
Medchemexpress LLC 5'-benzyl-1'-butyl-N-(naphthalen-2-ylsulfonyl)-6'-oxo-1',6'-dihydro-[2,3'-bipyridine]-4-carboxamide | 2139330-34-8 | 99.7% | 551.66 | C32H29N3O4S | 10MG
SDP
Catalog No. 5000237117
Encompass_Preferred
SERCA2a activator 1 is a small-molecule research compound that activates the sarco/endoplasmic reticulum Ca2+-dependent ATPase 2a (SERCA2a), attenuates phospholamban inhibition, and is used in preclinical studies of cardiac function and heart failure.
  • Activates SERCA2a and enhances Ca2+-dependent ATPase activity.
  • Attenuates phospholamban inhibition to improve systolic and diastolic function in models.
  • Suitable for preclinical cardiac function and heart failure research.
  • High purity (99.7%) with molecular weight 551.66.
  • Available as powder and as a 10 mM solution in DMSO, multiple package sizes.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).
Catalog No. 50-002-37117 Supplier Medchemexpress LLC Supplier No. HY12487310MG
SERCA2a activator 1 is a small-molecule research compound that activates the sarco/endoplasmic reticulum Ca2+-dependent ATPase 2a (SERCA2a), attenuates phospholamban inhibition, and is used in preclinical studies of cardiac function and heart failure.
  • Activates SERCA2a and enhances Ca2+-dependent ATPase activity.
  • Attenuates phospholamban inhibition to improve systolic and diastolic function in models.
  • Suitable for preclinical cardiac function and heart failure research.
  • High purity (99.7%) with molecular weight 551.66.
  • Available as powder and as a 10 mM solution in DMSO, multiple package sizes.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).