Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-benzyl-1'-butyl-N-(naphthalen-2-ylsulfonyl)-6'-oxo-1',6'-dihydro-[2,3'-bipyridine]-4-carboxamide | 2139330-34-8 | 99.5% | 551.66 g·mol⁻¹ | C32H29N3O4S | 5MG
SDP

Catalog No. 5000235385
Encompass_Preferred

SERCA2a activator 1 is a small-molecule activator of the sarco/endoplasmic reticulum Ca2+-dependent ATPase 2a (SERCA2a) used in preclinical cardiovascular research to attenuate phospholamban inhibition and enhance cardiac contractile function. Supplied for research use only.

  • Activates SERCA2a and attenuates phospholamban inhibition.
  • Used to enhance systolic and diastolic cardiac function in research models.
  • Molecular formula C32H29N3O4S.
  • Molecular weight 551.66 g·mol⁻¹.
  • Available in milligram quantities suitable for in vitro and in vivo studies.
  • Supplied for research use only; not for human or veterinary use.

Catalog No. 50-002-35385 Supplier Medchemexpress LLC Supplier No. HY1248735MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SERCA2a activator 1 is a small-molecule activator of the sarco/endoplasmic reticulum Ca2+-dependent ATPase 2a (SERCA2a) used in preclinical cardiovascular research to attenuate phospholamban inhibition and enhance cardiac contractile function. Supplied for research use only.

  • Activates SERCA2a and attenuates phospholamban inhibition.
  • Used to enhance systolic and diastolic cardiac function in research models.
  • Molecular formula C32H29N3O4S.
  • Molecular weight 551.66 g·mol⁻¹.
  • Available in milligram quantities suitable for in vitro and in vivo studies.
  • Supplied for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.