Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Benzyl-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. B308625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B308625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B308625G Supplier TCI America Supplier No. B308625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18489-25-3
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD00068730
InChI Key HHDRWGJJZGJSGZ-UHFFFAOYSA-N
Synonym 5-benzyl-1h-tetrazole, 5-benzyltetrazole, 1h-tetrazole, 5-phenylmethyl, 5-benzyl-2h-1,2,3,4-tetraazole, 5-benzyl-1h-1,2,3,4-tetrazole, 5-benzyl-2h-1,2,3,4-tetrazole, 5-phenylmethyltetrazole, maybridge1_003729, acmc-209em5, 5-phenylmethyl-1h-tetrazole
PubChem CID 223451
IUPAC Name 5-benzyl-2H-1,2,3,4-tetrazole
SMILES C(C1=NNN=N1)C1=CC=CC=C1

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 25 g
Formula Weight 160.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyl-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.