Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-benzyl-2-hydroxy-3-nitrobenzaldehyde | 328565-16-8 | MFCD00852947 | ≥95.0% | 257.24 g/mol | C14H11NO4 | 100MG
SDP

Catalog No. 5000239544
Encompass_Preferred

Col003 (5-benzyl-2-hydroxy-3-nitrobenzaldehyde; CAS 328565-16-8) is a small-molecule inhibitor of heat shock protein 47 (Hsp47) that competitively binds the collagen-binding site, disrupts Hsp47-collagen interaction, and inhibits collagen secretion. It is used for research into fibrosis and collagen-processing pathways.

  • Selective Hsp47 inhibitor with reported IC50 = 1.8 μM.
  • Disrupts collagen secretion by destabilizing the collagen triple helix.
  • Suitable for fibrosis and collagen pathway studies in vitro.
  • Molecular formula C14H11NO4; molecular weight 257.24 g/mol.
  • Soluble in DMSO (50 mg/mL); in vivo solubility reported 2.5 mg/mL.
  • Storage recommendations: solid at 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month) under nitrogen.

Catalog No. 50-002-39544 Supplier Medchemexpress LLC Supplier No. HY124817100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Col003 (5-benzyl-2-hydroxy-3-nitrobenzaldehyde; CAS 328565-16-8) is a small-molecule inhibitor of heat shock protein 47 (Hsp47) that competitively binds the collagen-binding site, disrupts Hsp47-collagen interaction, and inhibits collagen secretion. It is used for research into fibrosis and collagen-processing pathways.

  • Selective Hsp47 inhibitor with reported IC50 = 1.8 μM.
  • Disrupts collagen secretion by destabilizing the collagen triple helix.
  • Suitable for fibrosis and collagen pathway studies in vitro.
  • Molecular formula C14H11NO4; molecular weight 257.24 g/mol.
  • Soluble in DMSO (50 mg/mL); in vivo solubility reported 2.5 mg/mL.
  • Storage recommendations: solid at 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month) under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.