Learn More
Medchemexpress LLC 5-benzyl-2-hydroxy-3-nitrobenzaldehyde | 328565-16-8 | MFCD00852947 | 99.9% | 257.24 g·mol⁻¹ | C14H11NO4 | 50MG
SDP
Catalog No. 5000235589
Encompass_Preferred
Col003 (5-benzyl-2-hydroxy-3-nitrobenzaldehyde) is a selective small-molecule inhibitor of heat shock protein 47 (Hsp47) that competitively binds the collagen-binding site. It has a reported IC50 of 1.8 μM and is supplied as a high-purity research reagent for in vitro studies of collagen secretion and fibrosis-related pathways.
  • Selective Hsp47 inhibition (IC50 = 1.8 μM).
  • High purity suitable for biochemical assays (99.87%).
  • Molecular formula C14H11NO4, molecular weight 257.24 g·mol⁻¹.
  • Available in small pack sizes for research use, including 50 mg.
  • Used to investigate collagen secretion and fibrosis-related mechanisms.
Catalog No. 50-002-35589 Supplier Medchemexpress LLC Supplier No. HY12481750MG
Col003 (5-benzyl-2-hydroxy-3-nitrobenzaldehyde) is a selective small-molecule inhibitor of heat shock protein 47 (Hsp47) that competitively binds the collagen-binding site. It has a reported IC50 of 1.8 μM and is supplied as a high-purity research reagent for in vitro studies of collagen secretion and fibrosis-related pathways.
  • Selective Hsp47 inhibition (IC50 = 1.8 μM).
  • High purity suitable for biochemical assays (99.87%).
  • Molecular formula C14H11NO4, molecular weight 257.24 g·mol⁻¹.
  • Available in small pack sizes for research use, including 50 mg.
  • Used to investigate collagen secretion and fibrosis-related mechanisms.