Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Benzyloxy-DL-tryptophan 98.0+%, TCI America™
SDP

Catalog No. B0430100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0430100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0430100MG Supplier TCI America Supplier No. B0430100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1956-25-8
Molecular Formula C18H18N2O3
Molecular Weight (g/mol) 310.353
MDL Number MFCD00037968
InChI Key DFGNDJBYANKHIO-UHFFFAOYSA-N
Synonym H-DL-Trp(5-OBzl)-OH
PubChem CID 97908
IUPAC Name 2-amino-3-(5-phenylmethoxy-1H-indol-3-yl)propanoic acid
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3CC(C(=O)O)N

Specifications

Quantity 100 mg
Formula Weight 310.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyloxy-DL-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.