Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Benzyloxygramine | 1453-97-0 | 99.36% | 280.36 | 250 MG
SDP

Catalog No. 5000348786
Encompass_Preferred

5-Benzyloxygramine is an N protein PPI orthosteric stabilizer that demonstrates both antiviral and N-NTD protein-stabilizing activities. It stabilizes N-NTD dimers through simultaneous hydrophobic interactions with both partners, leading to abnormal N protein oligomerization, which has been confirmed in cells.

  • Acts as an antiviral and N-NTD protein-stabilizing agent
  • Stabilizes N-NTD dimers
  • Target: Antibiotic
  • Pathway: Anti-infection
  • Soluble in DMSO at 250 mg/mL
  • In vivo solubility ≥ 2.08 mg/mL in various solvent mixtures
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-48786 Supplier Medchemexpress LLC Supplier No. 250MGHY138694
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Benzyloxygramine is an N protein PPI orthosteric stabilizer that demonstrates both antiviral and N-NTD protein-stabilizing activities. It stabilizes N-NTD dimers through simultaneous hydrophobic interactions with both partners, leading to abnormal N protein oligomerization, which has been confirmed in cells.

  • Acts as an antiviral and N-NTD protein-stabilizing agent
  • Stabilizes N-NTD dimers
  • Target: Antibiotic
  • Pathway: Anti-infection
  • Soluble in DMSO at 250 mg/mL
  • In vivo solubility ≥ 2.08 mg/mL in various solvent mixtures
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.