Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Benzylthio)-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. B302025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B302025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B302025G Supplier TCI America Supplier No. B302025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21871-47-6
Molecular Formula C8H8N4S
Molecular Weight (g/mol) 192.24
MDL Number MFCD00068731
InChI Key GXGKKIPUFAHZIZ-UHFFFAOYSA-N
PubChem CID 323185
IUPAC Name 5-benzylsulfanyl-2H-tetrazole
SMILES C1=CC=C(C=C1)CSC2=NNN=N2

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 25 g
UN Number 1325
Formula Weight 192.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Benzylthio)-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.