Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bis(2-cyano-3-pyridylmethyl)amino-2-cyanopyridine, 97%, Thermo Scientific™

Catalog No. AAH5851503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5851503 1 g
AAH5851506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5851503 Supplier Thermo Scientific Chemicals Supplier No. H5851503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421263-82-2
Molecular Formula C20H13N7
Molecular Weight (g/mol) 351.373
MDL Number MFCD22682983
InChI Key MGYJYXRHCRJQQV-UHFFFAOYSA-N
Synonym 5-bis 2-cyano-3-pyridylmethyl amino-2-cyanopyridine, 5-bis 2-cyanopyridin-3-yl methyl amino pyridine-2-carbonitrile, 3-6-cyanopyridin-3-yl 2-cyanopyridin-3-yl methyl amino methyl pyridine-2-carbonitrile
PubChem CID 97030353
IUPAC Name 3-[[(6-cyanopyridin-3-yl)-[(2-cyanopyridin-3-yl)methyl]amino]methyl]pyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)CN(CC2=C(N=CC=C2)C#N)C3=CN=C(C=C3)C#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 351.37
Percent Purity 97%
Chemical Name or Material 5-Bis(2-cyano-3-pyridylmethyl)amino-2-cyanopyridine
missing translation for 'healthHazard1' H302+H312+H332-H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.