Learn More
Electron Microscopy Sciences 5 Block Embedding Mold-2 Part- 8 mm Body with 2 mm Cylindrical
SDP
Catalog No. 501902103
Encompass_Preferred
The block embedding mold features an innovative new design based on the widely used Ø8 mm size embedding mold. The unique two part design allows for easy removal of resin blocks, as well as incorporating many other advantages over currently available embedding products.
It has separate base and tube sections that clip together to form the familiar capsule shape. It allows for easy removal of resin blocks achieved by simply pulling the mold apart by hand.
It can produce blocks up to 20 mm in length with no razor blades or capsule presses are required. Each mold can be clipped to other molds, allowing for the creation of trays of desired size. Each cavity is numbered 1 < 5, whilst the end support stands have designated areas for writing indexing numbers or comments.
Catalog No. 50-190-2103 Supplier Electron Microscopy Sciences Supplier No. 6993430
The block embedding mold features an innovative new design based on the widely used Ø8 mm size embedding mold. The unique two part design allows for easy removal of resin blocks, as well as incorporating many other advantages over currently available embedding products.
It has separate base and tube sections that clip together to form the familiar capsule shape. It allows for easy removal of resin blocks achieved by simply pulling the mold apart by hand.
It can produce blocks up to 20 mm in length with no razor blades or capsule presses are required. Each mold can be clipped to other molds, allowing for the creation of trays of desired size. Each cavity is numbered 1 < 5, whilst the end support stands have designated areas for writing indexing numbers or comments.