Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-BOC-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine, 97%, Thermo Scientific™

Catalog No. AC450020010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450020010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450020010 Supplier Thermo Scientific Chemicals Supplier No. 450020010
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 230301-11-8
Molecular Formula C11H17N3O2
Molecular Weight (g/mol) 223.27
InChI Key BHYPERDTLBCHAE-UHFFFAOYSA-N
Synonym tert-butyl 6,7-dihydro-1h-pyrazolo 4,3-c pyridine-5 4h-carboxylate, 5-boc-1,4,6,7-tetrahydropyrazolo 4,3-c pyridine, tert-butyl 6,7-dihydro-2h-pyrazolo 4,3-c pyridine-5 4h-carboxylate, 5-boc-4,5,6,7-tetrahydropyrazolo 4,3-c pyridine, tert-butyl 2h,4h,5h,6h,7h-pyrazolo 4,3-c pyridine-5-carboxylate, tert-butyl 1h,4h,5h,6h,7h-pyrazolo 4,3-c pyridine-5-carboxylate, tert-butyl 2,4,6,7-tetrahydro-5h-pyrazolo-4,3-c pyridine-5-carboxylate, tert-butyl 2,4,6,7-tetrahydro-5h-pyrazolo 4,3-c pyridine-5-carboxylate, t-butyl 1h,4h,6h,7h-pyrazolo 4,3-c pyridine-5-carboxylate, tert-butyl 1h,4h,6h,7h-pyrazolo 4,3-c pyridine-5-carboxylate
PubChem CID 10398680
IUPAC Name tert-butyl 1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine-5-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2=C(C1)C=NN2

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 223.27
Percent Purity 97%
Chemical Name or Material 5-BOC-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.