Learn More
Medchemexpress LLC 5-(Boc-amino)-1-pentanol | 75178-90-4 | MFCD01862955 | ≥97.0% | 203.28 g/mol | C10H21NO3 | 10 G
SDP
Catalog No. 5000299763
Encompass_Preferred
5-(Boc-amino)-1-pentanol is a Boc-protected amino alcohol used as a versatile linker and building block in organic and medicinal chemistry. The tert-butoxycarbonyl (Boc) group protects the amine during multi-step syntheses, while the primary hydroxyl group enables subsequent functionalization and conjugation.
  • Boc-protected amine for orthogonal protection strategies.
  • Primary hydroxyl enables esterification and coupling reactions.
  • High purity suitable for synthesis (≥97%).
  • Available in multiple small-scale packages for research use.
  • Compact molecular weight facilitates incorporation into small molecules.
Catalog No. 50-002-99763 Supplier Medchemexpress LLC Supplier No. HYW03972910G
5-(Boc-amino)-1-pentanol is a Boc-protected amino alcohol used as a versatile linker and building block in organic and medicinal chemistry. The tert-butoxycarbonyl (Boc) group protects the amine during multi-step syntheses, while the primary hydroxyl group enables subsequent functionalization and conjugation.
  • Boc-protected amine for orthogonal protection strategies.
  • Primary hydroxyl enables esterification and coupling reactions.
  • High purity suitable for synthesis (≥97%).
  • Available in multiple small-scale packages for research use.
  • Compact molecular weight facilitates incorporation into small molecules.