Learn More
Medchemexpress LLC 5-(Boc-amino)-1-pentanol | 75178-90-4 | MFCD01862955 | ≥97.0% | 203.28 g/mol | C10H21NO3 | 25 G
SDP
Catalog No. 5000302904
Encompass_Preferred
5-(Boc-amino)-1-pentanol is a Boc-protected amino alcohol used as a versatile linker and intermediate in organic synthesis and medicinal chemistry. It features a tert-butoxycarbonyl (Boc) protected amine and a primary hydroxyl group that permit orthogonal functionalization - the hydroxyl can be derivatized or replaced, and the Boc group can be removed under mild acidic conditions to reveal the free amine.
  • Contains a Boc-protected amine and a primary hydroxyl.
  • Enables orthogonal functionalization for multi-step syntheses.
  • Hydroxyl group suitable for derivatization or conjugation reactions.
  • Boc group deprotects under mild acidic conditions to yield free amine.
  • High purity (≥97%) appropriate for research applications.
  • Solid material stable at room temperature with defined solvent storage conditions.
Catalog No. 50-003-02904 Supplier Medchemexpress LLC Supplier No. 25GHYW039729
5-(Boc-amino)-1-pentanol is a Boc-protected amino alcohol used as a versatile linker and intermediate in organic synthesis and medicinal chemistry. It features a tert-butoxycarbonyl (Boc) protected amine and a primary hydroxyl group that permit orthogonal functionalization - the hydroxyl can be derivatized or replaced, and the Boc group can be removed under mild acidic conditions to reveal the free amine.
  • Contains a Boc-protected amine and a primary hydroxyl.
  • Enables orthogonal functionalization for multi-step syntheses.
  • Hydroxyl group suitable for derivatization or conjugation reactions.
  • Boc group deprotects under mild acidic conditions to yield free amine.
  • High purity (≥97%) appropriate for research applications.
  • Solid material stable at room temperature with defined solvent storage conditions.