Learn More
Medchemexpress LLC 5-(Boc-amino)pentanoic acid | 27219-07-4 | MFCD00076903 | 99.9% | 217.26 g·mol-1 | C10H19NO4 | 5 G
SDP
Catalog No. 5000304048
Encompass_Preferred
Boc-NH-C4-acid is a Boc-protected 5-aminovaleric acid used as an alkyl linker in PROTAC and medicinal chemistry applications. It contains a Boc-protected amine and a terminal carboxylic acid, allowing orthogonal coupling and downstream deprotection. Molecular formula C10H19NO4; molecular weight 217.26 g·mol-1. High nominal purity supports synthetic workflows.
  • Boc-protected amine enables temporary protection during multi-step synthesis.
  • Terminal carboxylic acid allows efficient amide coupling reactions.
  • Suitable for PROTAC linker assembly and linker optimization studies.
  • High nominal purity (99.85%) for reliable reproducibility.
  • Available in bench-scale quantities for research use.
Catalog No. 50-003-04048 Supplier Medchemexpress LLC Supplier No. HYW0140995G
Boc-NH-C4-acid is a Boc-protected 5-aminovaleric acid used as an alkyl linker in PROTAC and medicinal chemistry applications. It contains a Boc-protected amine and a terminal carboxylic acid, allowing orthogonal coupling and downstream deprotection. Molecular formula C10H19NO4; molecular weight 217.26 g·mol-1. High nominal purity supports synthetic workflows.
  • Boc-protected amine enables temporary protection during multi-step synthesis.
  • Terminal carboxylic acid allows efficient amide coupling reactions.
  • Suitable for PROTAC linker assembly and linker optimization studies.
  • High nominal purity (99.85%) for reliable reproducibility.
  • Available in bench-scale quantities for research use.