Learn More
Medchemexpress LLC HY-15910 100mg Medchemexpress, 5-BrdU CAS:59-14-3 Purity:>98%
SDP
Catalog No. 502029181
Encompass_Preferred
Medchemexpress, HY-15910 100mg 5-BrdU CAS:59-14-3 5-BrdU (BrdU) is a nucleoside analog that competes with thymidine for incorporation into DNA. 5-BrdU is commonly used in the detection of proliferating cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9181 Supplier Medchemexpress LLC Supplier No. HY15910100MG
Medchemexpress, HY-15910 100mg 5-BrdU CAS:59-14-3 5-BrdU (BrdU) is a nucleoside analog that competes with thymidine for incorporation into DNA. 5-BrdU is commonly used in the detection of proliferating cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.