Learn More
Medchemexpress LLC HY-15910 1g , 5-BrdU CAS:59-14-3 Purity:>98%
SDP
Catalog No. 502259542
Encompass_Preferred
HY-15910 1g 5-BrdU CAS: 59-14-3 5-BrdU (BrdU) is a nucleoside analog that competes with thymidine for incorporation into DNA. 5-BrdU is commonly used in the detection of proliferating cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9542 Supplier Medchemexpress LLC Supplier No. HY159101G
HY-15910 1g 5-BrdU CAS: 59-14-3 5-BrdU (BrdU) is a nucleoside analog that competes with thymidine for incorporation into DNA. 5-BrdU is commonly used in the detection of proliferating cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.