Learn More
Apexbio Technology LLC 5-BrdU(Synonyms: Bromodeoxyuridine, BrdU, 5-Bromo-2'-deoxyuridine, 5-Bromodeoxyuridine), 1g, CAS: 59-14-3.
SDP
Catalog No. 501151755
Encompass_Preferred
5-Bromo-2 -deoxyuridine (5-BrdU CAS 59-14-3) is a thymidine analog that integrates into DNA during S-phase of cell division Its incorporation facilitates immunodetection via specific monoclonal or polyclonal antibodies In vitro studies show that early application of 5-BrdU to Nicotiana glauca pith explants inhibits cellular proliferation an effect reversed by addition of thymidine or deoxycytidine In vivo BrdU immunostaining identifies proliferating neuronal cells in mouse central nervous system tissues paralleling earlier thymidine labeling studies Thus 5-BrdU serves as a molecular tool for analyzing cell cycle progression and neurogenesis in biomedical research
Catalog No. 50-115-1755 Supplier Apexbio Technology LLC Supplier No. B35891000
5-Bromo-2 -deoxyuridine (5-BrdU CAS 59-14-3) is a thymidine analog that integrates into DNA during S-phase of cell division Its incorporation facilitates immunodetection via specific monoclonal or polyclonal antibodies In vitro studies show that early application of 5-BrdU to Nicotiana glauca pith explants inhibits cellular proliferation an effect reversed by addition of thymidine or deoxycytidine In vivo BrdU immunostaining identifies proliferating neuronal cells in mouse central nervous system tissues paralleling earlier thymidine labeling studies Thus 5-BrdU serves as a molecular tool for analyzing cell cycle progression and neurogenesis in biomedical research